accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3POOCH3OCH3

CAS RN 17145-91-4

IUPAC Name: ethyl 2-(diethoxyphosphoryl)butanoate
Synonyms: ethyl 2-(diethoxyphosphoryl)butanoate
Molecular Weight (g/mol): 252.25
Molecular Formula: C10H21O5P
InChi Key: GYUCVQSNZFRDRL-UHFFFAOYNA-N
SMILES: CCOC(=O)C(CC)P(=O)(OCC)OCC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)