accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CCl

CAS RN 17146-09-7

IUPAC Name: chlorodimethyl(3-phenylpropyl)silane
Synonyms: chlorodimethyl(3-phenylpropyl)silane
Molecular Weight (g/mol): 212.79
Molecular Formula: C11H17ClSi
InChi Key: ASSMBLOISZSMMP-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CCCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results