accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17153-21-8

IUPAC Name: 3-methyl-5-phenyl-1,2-oxazole-4-carboxylic acid
Synonyms: 3-methyl-5-phenyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 203.2
Molecular Formula: C11H9NO3
InChi Key: GLNQCTGGLIXRRJ-UHFFFAOYSA-N
SMILES: CC1=NOC(=C1C(O)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results