accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17159-80-7

IUPAC Name: ethyl 4-hydroxycyclohexane-1-carboxylate
Synonyms: ethyl 4-hydroxycyclohexane-1-carboxylate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: BZKQJSLASWRDNE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCC(O)CC1
Boiling Point 
  • (3)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results