accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 171611-11-3

IUPAC Name: lithium(1+) bis(fluorosulfonyl)azanide
Synonyms: lithium(1+) bis(fluorosulfonyl)azanide
Molecular Weight (g/mol): 187.06
Molecular Formula: F2LiNO4S2
InChi Key: VDVLPSWVDYJFRW-UHFFFAOYSA-N
SMILES: [Li+].FS(=O)(=O)[N-]S(F)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results