accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 171661-55-5

IUPAC Name: 1-(5-methylthiophen-2-yl)methanamine hydrochloride
Synonyms: 1-(5-methylthiophen-2-yl)methanamine hydrochloride
Molecular Weight (g/mol): 163.66
Molecular Formula: C6H10ClNS
InChi Key: COTMVCYERKXKDX-UHFFFAOYSA-N
SMILES: Cl.CC1=CC=C(CN)S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results