accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1718-52-1

IUPAC Name: (²H₁₀)pyrene
Synonyms: (²H₁₀)pyrene
Molecular Weight (g/mol): 212.32
Molecular Formula: C16H10
InChi Key: BBEAQIROQSPTKN-LHNTUAQVSA-N
SMILES: [2H]C1=C([2H])C2=C([2H])C([2H])=C3C([2H])=C([2H])C([2H])=C4C([2H])=C([2H])C(=C1[2H])C2=C34

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results