accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1718-53-2

IUPAC Name: tetraphene
Synonyms: benz[a]anthracene naphthanthracene 1,2-benz(a)anthracene benzanthracene benz(a)anthracene benzo(a)anthracene benzoanthracene 1,2-benzoanthracene 2,3-benzphenanthrene benzo(b)phenanthrene
Molecular Weight (g/mol): 228.29
Molecular Formula: C18H12
InChi Key: DXBHBZVCASKNBY-UHFFFAOYSA-N
SMILES: C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results