accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHZnBr

CAS RN 171860-65-4

IUPAC Name: adamantan-2-ide; bromozincylium
Synonyms: adamantan-2-ide; bromozincylium
Molecular Weight (g/mol): 280.51
Molecular Formula: C10H15BrZn
InChi Key: UKVCFEOZGIQMNI-UHFFFAOYSA-M
SMILES: [Zn+]Br.C1C2CC3CC1CC(C2)[CH-]3

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)