accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CHH3CZnBr

CAS RN 171860-66-5

IUPAC Name: bromozincylium; butan-2-ide
Synonyms: bromozincylium; butan-2-ide
Molecular Weight (g/mol): 202.4
Molecular Formula: C4H9BrZn
InChi Key: DATGUFISTUEOBS-UHFFFAOYSA-M
SMILES: [Zn+]Br.CC[CH-]C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)