accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 171887-03-9

IUPAC Name: N-(2-amino-4,6-dichloropyrimidin-5-yl)formamide
Synonyms: N-(2-amino-4,6-dichloropyrimidin-5-yl)formamide
Molecular Weight (g/mol): 207.01
Molecular Formula: C5H4Cl2N4O
InChi Key: XYWHZUCZNRMJGO-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=C(NC=O)C(Cl)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results