accessibility menu, dialog, popup

UserName

Viewing profile as user:
2H2H2H2H2H2H2H2H2H2H2H2H

CAS RN 1719-03-5

IUPAC Name: (²H₁₂)chrysene
Synonyms: (²H₁₂)chrysene
Molecular Weight (g/mol): 240.37
Molecular Formula: C18H12
InChi Key: WDECIBYCCFPHNR-AQZSQYOVSA-N
SMILES: [2H]C1=C([2H])C([2H])=C2C(C([2H])=C([2H])C3=C2C([2H])=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C32)=C1[2H]
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results