accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17194-00-2

IUPAC Name: barium(2+) dihydroxide
Synonyms: barium hydroxide bariumhydroxid aetzbaryt Ba(OH)2 barium hydroxide lime caustic baryta
Molecular Weight (g/mol): 171.34
Molecular Formula: BaH2O2
InChi Key: RQPZNWPYLFFXCP-UHFFFAOYSA-L
SMILES: [OH-].[OH-].[Ba++]
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)