accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17199-29-0

IUPAC Name: (2S)-2-hydroxy-2-phenylacetic acid
Synonyms: (S)-mandelic acid (S)-mandelsaeure
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: IWYDHOAUDWTVEP-ZETCQYMHSA-N
SMILES: O[C@H](C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
  • (2)
Quantity 
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (2)
  • (5)
  • (7)
  • (2)
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)