accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONH3C

CAS RN 1721-26-2

IUPAC Name: ethyl 2-methylpyridine-3-carboxylate
Synonyms: ethyl 2-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: TZORNSWZUZDUCU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CN=C1C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)