CAS RN 172100-44-6
CAS RN 172100-44-6
IUPAC Name:
5-bromo-2-(5-bromo-4-nonyl-1,3-thiazol-2-yl)-4-nonyl-1,3-thiazole
Synonyms:
5-bromo-2-(5-bromo-4-nonyl-1,3-thiazol-2-yl)-4-nonyl-1,3-thiazole
Molecular Weight (g/mol):
578.51
Molecular Formula:
C24H38Br2N2S2
InChi Key:
NVYFBJQBTRFBCJ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCC1=C(Br)SC(=N1)C1=NC(CCCCCCCCC)=C(Br)S1
1
–
1
of
1
results
1
5,5'-Dibromo-4,4'-dinonyl-2,2'-bithiazole 97.0+%, TCI America™
CAS: 172100-44-6 Molecular Formula: C24H38Br2N2S2 Molecular Weight (g/mol): 578.51 InChI Key: NVYFBJQBTRFBCJ-UHFFFAOYSA-N PubChem CID: 19437292 IUPAC Name: 5-bromo-2-(5-bromo-4-nonyl-1,3-thiazol-2-yl)-4-nonyl-1,3-thiazole SMILES: CCCCCCCCCC1=C(SC(=N1)C2=NC(=C(S2)Br)CCCCCCCCC)Br