accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1722-62-9

IUPAC Name: hydrogen N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide chloride
Synonyms: hydrogen mepivacaine chloride
Molecular Weight (g/mol): 282.81
Molecular Formula: C15H23ClN2O
InChi Key: RETIMRUQNCDCQB-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN1CCCCC1C(=O)NC1=C(C)C=CC=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results