accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNHClO

CAS RN 17228-70-5

IUPAC Name: 3,5-dichloro-1,4-dihydropyridin-4-one
Synonyms: 3,5-dichloro-1H-pyridin-4-one
Molecular Weight (g/mol): 163.99
Molecular Formula: C5H3Cl2NO
InChi Key: YKCQWIYRLMNGMO-UHFFFAOYSA-N
SMILES: ClC1=CNC=C(Cl)C1=O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results