accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17228-99-8

IUPAC Name: 2-(4-methylphenyl)-1,3-thiazole-4-carboxylate
Synonyms: 2-(4-methylphenyl)-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 218.25
Molecular Formula: C11H8NO2S
InChi Key: MTVWIZYQYPRZGZ-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)C1=NC(=CS1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results