accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 172285-72-2

IUPAC Name: 9,9',10,10'-tetraphenyl-2,2'-bianthracene
Synonyms: 9,9',10,10'-tetraphenyl-2,2'-bianthracene
Molecular Weight (g/mol): 658.84
Molecular Formula: C52H34
InChi Key: BHPFDLWDNJSMOS-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC(=CC=C12)C1=CC=C2C(=C1)C(C1=CC=CC=C1)=C1C=CC=CC1=C2C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results