accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClO

CAS RN 17229-17-3

IUPAC Name: [(2-chloroethoxy)methyl]benzene
Synonyms: [(2-chloroethoxy)methyl]benzene
Molecular Weight (g/mol): 170.64
Molecular Formula: C9H11ClO
InChi Key: UPIMXDQREQJWMR-UHFFFAOYSA-N
SMILES: ClCCOCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results