accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHN

CAS RN 1723-00-8

IUPAC Name: piperidine-2-carboxylic acid
Synonyms: (+,-)-pipecolic acid pipecolic acid, (+,-)-isomer pipecolic acid free base pipecolic acid, (S)-isomer DL-pipecolate pipecolic acid, (r)- pipecolic acid, (s)- (R)-isomer pipecolic acid (s)-isomer pipecolic acid L-(-)-pipecolate
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: HXEACLLIILLPRG-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCCN1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)