accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17250-25-8

IUPAC Name: methanidylidyneoxidanium tris(triphenylphosphane) iridium hydride
Synonyms: carbon monoxide tris(triphenylphosphine) iridium hydride
Molecular Weight (g/mol): 1008.11
Molecular Formula: C55H46IrOP3
InChi Key: IOPVMNYABUEALT-UHFFFAOYSA-N
SMILES: [IrH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results