CAS RN 172507-33-4
CAS RN 172507-33-4
IUPAC Name:
4-(thiophen-3-yl)benzonitrile
Synonyms:
4-(thiophen-3-yl)benzonitrile
Molecular Weight (g/mol):
185.24
Molecular Formula:
C11H7NS
InChi Key:
GRAMVGQZHXAXHP-UHFFFAOYSA-N
SMILES:
N#CC1=CC=C(C=C1)C1=CSC=C1
1
–
2
of
2
results
1
4-Thien-3-ylbenzonitrile, ≥97%, Thermo Scientific™
CAS: 172507-33-4 Molecular Formula: C11H7NS Molecular Weight (g/mol): 185.244 MDL Number: MFCD00115000 InChI Key: GRAMVGQZHXAXHP-UHFFFAOYSA-N Synonym: 4-3-thienyl benzonitrile,4-thiophen-3-yl benzonitrile,4-thien-3-ylbenzonitrile,benzonitrile, 4-3-thienyl,4-3-thienyl benzenecarbonitrile PubChem CID: 4130333 IUPAC Name: 4-thiophen-3-ylbenzonitrile SMILES: C1=CC(=CC=C1C#N)C2=CSC=C2
2
Sigma Aldrich 4-(3-Thienyl)benzonitrile
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.