accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 172531-37-2

IUPAC Name: 2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}acetic acid
Synonyms: {2-[2-(2-azidoethoxy)ethoxy]ethoxy}acetic acid
Molecular Weight (g/mol): 233.22
Molecular Formula: C8H15N3O5
InChi Key: GIXBCECBLAEYKA-UHFFFAOYSA-N
SMILES: OC(=O)COCCOCCOCCN=[N+]=[N-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results