CAS RN 172656-11-0
CAS RN 172656-11-0
IUPAC Name:
18-bromooctadec-1-ene
Synonyms:
18-bromooctadec-1-ene
Molecular Weight (g/mol):
331.38
Molecular Formula:
C18H35Br
InChi Key:
HGBDROSSUKYLIG-UHFFFAOYSA-N
SMILES:
BrCCCCCCCCCCCCCCCCC=C
1
–
1
of
1
results
1
18-Bromo-1-octadecene 93.0+%, TCI America™
CAS: 172656-11-0 Molecular Formula: C18H35Br Molecular Weight (g/mol): 331.382 InChI Key: HGBDROSSUKYLIG-UHFFFAOYSA-N Synonym: 17-Octadecenyl Bromide PubChem CID: 15192957 IUPAC Name: 18-bromooctadec-1-ene SMILES: C=CCCCCCCCCCCCCCCCCBr