accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOSCl

CAS RN 17266-30-7

IUPAC Name: 2-(5-chloro-1-benzothiophen-3-yl)acetic acid
Synonyms: (5-chloro-1-benzothiophen-3-yl)acetic acid
Molecular Weight (g/mol): 226.67
Molecular Formula: C10H7ClO2S
InChi Key: QQKKTOPRRGBBCT-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CSC2=CC=C(Cl)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results