accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(S)N

CAS RN 172695-25-9

IUPAC Name: tert-butyl N-[(2S)-1-cyano-3-phenylpropan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-cyano-3-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol): 260.34
Molecular Formula: C15H20N2O2
InChi Key: BEJMQXQXIQDKCZ-CYBMUJFWSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC#N)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results