accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17273-79-9

IUPAC Name: sodium naphthalene-2-carboxylate
Synonyms: sodium 2-naphthoate
Molecular Weight (g/mol): 194.17
Molecular Formula: C11H7NaO2
InChi Key: WUJXROKYJRAVIO-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results