accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)NHCH3H3C

CAS RN 17279-31-1

IUPAC Name: dimethyl[(1S)-1-phenylethyl]azanium
Synonyms: dimethyl[(1S)-1-phenylethyl]azanium
Molecular Weight (g/mol): 150.24
Molecular Formula: C10H16N
InChi Key: BVURNMLGDQYNAF-VIFPVBQESA-O
SMILES: C[C@H]([NH+](C)C)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)