accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17281-59-3

IUPAC Name: 1-(cyanomethyl)pyridin-1-ium chloride
Synonyms: 1-(cyanomethyl)pyridin-1-ium chloride
Molecular Weight (g/mol): 154.6
Molecular Formula: C7H7ClN2
InChi Key: HEJOROWXIWLCMS-UHFFFAOYSA-M
SMILES: [Cl-].N#CC[N+]1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)