accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SOOOH

CAS RN 17286-26-9

IUPAC Name: 5-hydroxynaphthalene-1-sulfonamide
Synonyms: 5-hydroxynaphthalene-1-sulfonamide
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: NFVBVKHGDDDCEA-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=C2C=CC=C(O)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results