accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OH(R)CH3OH

CAS RN 17299-07-9

IUPAC Name: (2R,5R)-hexane-2,5-diol
Synonyms: (2R,5R)-hexane-2,5-diol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: OHMBHFSEKCCCBW-PHDIDXHHSA-N
SMILES: C[C@@H](O)CC[C@@H](C)O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)