accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3

CAS RN 1730-37-6

IUPAC Name: 1-methyl-9H-fluorene
Synonyms: 1-methylfluorene
Molecular Weight (g/mol): 180.25
Molecular Formula: C14H12
InChi Key: GKEUODMJRFDLJY-UHFFFAOYSA-N
SMILES: CC1=CC=CC2=C1CC1=CC=CC=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results