accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17303-83-2

IUPAC Name: (3-formylthiophen-2-yl)boronic acid
Synonyms: 3-formylthiophen-2-ylboronic acid
Molecular Weight (g/mol): 155.96
Molecular Formula: C5H5BO3S
InChi Key: HYXMHAHVUFTVFZ-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=O)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results