accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNCH3H3CCH3H3CCH3H3CCl

CAS RN 173035-10-4

IUPAC Name: 1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium chloride
Synonyms: 1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-1λ⁵-imidazol-1-ylium chloride
Molecular Weight (g/mol): 342.91
Molecular Formula: C21H27ClN2
InChi Key: COGMCBFILULEOS-UHFFFAOYSA-M
SMILES: [Cl-].CC1=CC(C)=C(N2CC[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)