accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNBrBr

CAS RN 173063-52-0

IUPAC Name: 3,6-dibromo-9-(2-ethylhexyl)-9H-carbazole
Synonyms: 3,6-dibromo-9-(2-ethylhexyl)carbazole
Molecular Weight (g/mol): 437.22
Molecular Formula: C20H23Br2N
InChi Key: ZDFZWZGIGKUBRA-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results