accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOO

CAS RN 17309-39-6

IUPAC Name: [3,3'-bioxolane]-2,2',5,5'-tetrone
Synonyms: [3,3'-bioxolane]-2,2',5,5'-tetrone
Molecular Weight (g/mol): 198.13
Molecular Formula: C8H6O6
InChi Key: OLQWMCSSZKNOLQ-UHFFFAOYNA-N
SMILES: O=C1CC(C2CC(=O)OC2=O)C(=O)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results