accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1731-79-9

H3COOOOCH3

CAS RN 1731-79-9

IUPAC Name: 1,12-dimethyl dodecanedioate
Synonyms: dodecanedioic acid, dimethyl ester
Molecular Weight (g/mol): 258.36
Molecular Formula: C14H26O4
InChi Key: IZMOTZDBVPMOFE-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCCCC(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results