accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 1732-10-1

IUPAC Name: 1,9-dimethyl nonanedioate
Synonyms: 1,9-dimethyl nonanedioate
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: DRUKNYVQGHETPO-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
Percent Purity 
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)