accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3NCl

CAS RN 17321-77-6

IUPAC Name: hydrogen (3-{5-chloro-2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
Synonyms: hydrogen clomipramine chloride
Molecular Weight (g/mol): 351.32
Molecular Formula: C19H24Cl2N2
InChi Key: WIMWMKZEIBHDTH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=C(Cl)C=C12
Molecular Weight (g/mol) 
  • (2)
  • (5)
Physical Form 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
  • (5)
Percent Purity 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (5)