accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHN

CAS RN 17325-26-7

IUPAC Name: methyl 1H-imidazole-5-carboxylate
Synonyms: methyl 3H-imidazole-4-carboxylate
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: DVLGIQNHKLWSRU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=CN1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)