accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17325-85-8

IUPAC Name: (2S)-3-(benzyloxy)propane-1,2-diol
Synonyms: (2S)-3-(benzyloxy)propane-1,2-diol
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: LWCIBYRXSHRIAP-JTQLQIEISA-N
SMILES: OC[C@H](O)COCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results