accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 173282-60-5

IUPAC Name: 6-(morpholin-4-yl)pyridine-3-carbaldehyde
Synonyms: 6-(morpholin-4-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: VZEANTSDLFVWCK-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results