accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1733-88-6

IUPAC Name: potassium phenyl sulfate
Synonyms: potassium phenyl sulfate
Molecular Weight (g/mol): 212.26
Molecular Formula: C6H5KO4S
InChi Key: NOUFXYHWAWIDNT-UHFFFAOYSA-M
SMILES: [K+].[O-]S(=O)(=O)OC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results