accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1733-89-7

IUPAC Name: potassium naphthalen-2-yl sulfate
Synonyms: potassium naphthalen-2-yl sulfate
Molecular Weight (g/mol): 262.32
Molecular Formula: C10H7KO4S
InChi Key: SKROFUUKJBLLKJ-UHFFFAOYSA-M
SMILES: [K+].[O-]S(=O)(=O)OC1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results