accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOOOONH2NHNH2(S)(S)(S)(S)CH3H3CCH3H3CCH3H3CH3CCH3

CAS RN 173334-57-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results