CAS RN 173336-76-0
IUPAC Name:
4-bromo-1-methoxy-2-(3-methoxypropoxy)benzene
Synonyms:
4-bromo-1-methoxy-2-(3-methoxypropoxy)benzene
Molecular Weight (g/mol):
275.14
Molecular Formula:
C11H15BrO3
InChi Key:
JFSCCLJKKCRXSS-UHFFFAOYSA-N
SMILES:
COCCCOC1=C(OC)C=CC(Br)=C1
1
–
1
of
1
results
1
4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene 98.0+%, TCI America™
CAS: 173336-76-0 Molecular Formula: C11H15BrO3 Molecular Weight (g/mol): 275.142 MDL Number: MFCD14582816 InChI Key: JFSCCLJKKCRXSS-UHFFFAOYSA-N PubChem CID: 21458711 IUPAC Name: 4-bromo-1-methoxy-2-(3-methoxypropoxy)benzene SMILES: COCCCOC1=C(C=CC(=C1)Br)OC