accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17339-60-5

IUPAC Name: (2-{[2-(dimethylazaniumyl)ethyl]disulfanyl}ethyl)dimethylazanium dichloride
Synonyms: (2-{[2-(dimethylammonio)ethyl]disulfanyl}ethyl)dimethylazanium dichloride
Molecular Weight (g/mol): 281.3
Molecular Formula: C8H22Cl2N2S2
InChi Key: OJNVDODUOWHJPK-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].C[NH+](C)CCSSCC[NH+](C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results