accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NFNH2

CAS RN 173435-32-0

IUPAC Name: 2-fluoro-4-methylpyridin-3-amine
Synonyms: 2-fluoro-4-methylpyridin-3-amine
Molecular Weight (g/mol): 126.13
Molecular Formula: C6H7FN2
InChi Key: ZKQQKPXTKXTEQR-UHFFFAOYSA-N
SMILES: CC1=C(N)C(F)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results